2-Iodo-6-methylpyridine 98%

Supplier: Ambeed
Warning

A100612-100MG A100612-250MG A100612-5G
103795-536EA 23 USD
103795-536 103821-226 76780-392 76780-394 77296-778
2-Iodo-6-methylpyridine 98%
2-Iodo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆IN
Boiling Pt: 222.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11044364
CAS Number: 62674-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR