2-Iodo-6-methoxypyridine 97%

Supplier: Ambeed
Warning

A135445-100MG A135445-250MG A135445-1G
103802-156EA 21 USD
103802-156 77010-114 77010-116 77314-982
2-Iodo-6-methoxypyridine 97%
2-Iodo-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.0200 g/mol
Boiling Pt: 248.4 °C (760 mm Hg)
Melting Pt: 39…40 °C
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07366733
CAS Number: 182275-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR