2-Iodo-5-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A394194-1G A394194-5G A394194-10G A394194-250MG
103826-940EA 18 USD
103826-940 103837-978 76841-386 76841-388 77264-444 77264-446
2-Iodo-5-nitrobenzoic acid 98%
2-Iodo-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄INO₄
Boiling Pt: 394.2 °C (760 mm Hg)
Flash Pt: 192.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00234256
CAS Number: 19230-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR