6-Iodo-m-toluic acid 98%

Supplier: Ambeed
Warning

A383582-25G A383582-100G A383582-10G A383582-500G A383582-5G
103837-780EA 5 USD
103837-780 76841-112 77264-262 77264-264 77264-266 77306-944
6-Iodo-m-toluic acid 98%
6-Iodo-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 321.4 °C (760 mm Hg)
Melting Pt: 122…124 °C
Flash Pt: 148.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079765
CAS Number: 52548-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR