2-Iodo-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A319622-1G A319622-5G A319622-250MG A319622-10G A319622-25G
103825-208EA 29 USD
103825-208 103836-140 76838-990 76838-992 77032-754 77668-420 77872-449
2-Iodo-5-(trifluoromethyl)benzoic acid 97%
2-Iodo-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃IO₂
Melting Pt: 171…172 °C
Flash Pt: 136.08 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11849931
CAS Number: 702641-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR