2-Iodo-5-(trifluoromethyl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-iodobenzotrifluoride

A270472-1G A270472-10G A270472-250MG A270472-5G
103813-002EA 30 USD
103813-002 103852-906 77092-676 77276-764 77276-766 77305-054
2-Iodo-5-(trifluoromethyl)aniline 95%
2-Iodo-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃IN
MW: 287.0210 g/mol
Boiling Pt: 249.7 °C (760 mm Hg)
Flash Pt: 101.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972658
CAS Number: 105202-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR