2-Iodo-4-methylphenol 95%

Supplier: Ambeed
Warning

A112071-10G A112071-25G A112071-1G A112071-5G A112071-250MG
103797-948EA 11 USD
103797-948 103823-782 76982-780 76982-782 77310-588 77310-590 77889-772
2-Iodo-4-methylphenol 95%
2-Iodo-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇IO
Boiling Pt: 208.9±20.0 °C (760 mm Hg)
Melting Pt: 35 °C
Flash Pt: 80.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00189405
CAS Number: 16188-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR