2-Iodo-p-toluidine 98%
Supplier: Ambeed
|
Warning
|
A122989-100G
A122989-10G
A122989-25G
A122989-1G
A122989-5G
103800-320EA
9
USD
103800-320
76766-668
76766-670
76766-672
77297-910
77297-912
2-Iodo-p-toluidine 98%
2-Iodo-p-toluidine
Precaución: for Research Use Only
Formula:
C₇H₈IN Boiling Pt: 268.7 °C (760 mm Hg) Melting Pt: 34…37 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03407391 CAS Number: 29289-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...