3-Iodo-4-nitroanisole 96%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-4-methoxy-1-nitrobenzene

A770151-1G A770151-25G A770151-5G A770151-10G
103840-932EA 35 USD
103840-932 76845-176 76845-178 76845-180 77146-374
3-Iodo-4-nitroanisole 96%
3-Iodo-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆INO₃
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD01320682
CAS Number: 214279-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR