4-Hydroxy-3-iodobenzotrifluoride 98%

Supplier: Ambeed
Warning

A214651-100MG A214651-250MG A214651-1G A214651-5G
103818-424EA 18 USD
103818-424 103849-128 77030-620 77030-622 77303-360 77303-362
4-Hydroxy-3-iodobenzotrifluoride 98%
4-Hydroxy-3-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₄F₃IO
Storage Temperature: Refrigerator
MDL Number: MFCD12922960
CAS Number: 463976-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR