2-Iodo-m-toluic acid 97%

Supplier: Ambeed
Warning

A139982-10G A139982-1G A139982-25G A139982-5G
103802-808EA 10 USD
103802-808 77258-752 77299-104 77299-106 77315-918
2-Iodo-m-toluic acid 97%
2-Iodo-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 323.5 °C (760 mm Hg)
Melting Pt: 150…153 °C
Flash Pt: 149.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00079764
CAS Number: 108078-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR