2-Iodo-1-methylimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-1-methyl-1H-imidazole

A254101-1G A254101-5G A254101-100MG A254101-250MG
103812-122EA 54 USD
103812-122 103851-976 77091-414 77091-416 77262-080 77262-082
2-Iodo-1-methylimidazole 97%
2-Iodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
MW: 208 g/mol
Boiling Pt: 263.4 °C (760 mm Hg)
Melting Pt: 87 °C
Flash Pt: 113.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00661606
CAS Number: 37067-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR