2-Iodo-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A289921-250MG A289921-1G
103813-854EA 84 USD
103813-854 103834-994 76816-560 76837-576
2-Iodo-1H-pyrrolo[2,3-b]pyridine 95%
2-Iodo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD16875827
CAS Number: 1227270-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR