2-Iodo-m-xylene 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dimethyl-2-iodobenzene, 2-Iodo-1,3-dimethylbenzene, 2,6-Dimethyliodobenzene, 2,6-Dimethyl-1-iodobenzene

A197260-1G A197260-25G A197260-5G A197260-100G A197260-10G
103816-904EA 7 USD
103816-904 103847-526 77085-514 77085-516 77085-518 77139-266 77139-268
2-Iodo-m-xylene 98%
2-Iodo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 223…224 °C (760 mm Hg)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00019012
CAS Number: 608-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR