2-Iodobiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-1,1'-biphenyl

A273054-100G A273054-10G A273054-25G A273054-5G A273054-1G
103813-124EA 5 USD
103813-124 103853-040 76723-084 76723-086 77092-834 77092-836 77305-120
2-Iodobiphenyl 98%
2-Iodobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉I
Boiling Pt: 331.7 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039396
CAS Number: 2113-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR