2-Hydroxyphenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

A201420-1G A201420-25G A201420-5G A201420-100G A201420-10G
103817-290EA 6 USD
103817-290 103847-928 77086-002 77086-004 77086-006 77274-858 77274-860
2-Hydroxyphenylboronic acid 98%
2-Hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Boiling Pt: 327.3 °C (760 mm Hg)
Melting Pt: 185 °C
Flash Pt: 151.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01074581
CAS Number: 89466-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR