2-Hydroxymethyl-3-methylpyridine 95%

Supplier: Ambeed
Warning

A222004-1G A222004-25G A222004-5G A222004-250MG
103818-960EA 13 USD
103818-960 103849-836 77088-430 77088-432 77088-434 77112-778
2-Hydroxymethyl-3-methylpyridine 95%
2-Hydroxymethyl-3-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
Boiling Pt: 236.8 °C (760 mm Hg)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD02684206
CAS Number: 63071-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR