Salicylhydrazide 97% (by HPLC)
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxybenzoic acid hydrazide, Salicylohydrazide, 2-Hydroxybenzene-1-carbohydrazide, Salicylic acid hydrazide, 2-Hydroxybenzohydrazide, 2-Hydroxybenzhydrazide, 2-Hydroxybenzoic hydrazide
A213806-25G
A213806-100G
A213806-10G
103849-050EA
8
USD
103849-050
77087-556
77192-722
77192-724
Salicylhydrazide 97% (by HPLC)
Salicylhydrazide
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 147…150 °C Storage Temperature: Freezer |
MDL Number:
MFCD00007599 CAS Number: 936-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...