2-Hydroxybenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Benzimidazolinone, 2(1H)-Benzimidazolone, 2-Hydroxy-1H-benzimidazole

A109011-100G A109011-10G A109011-25G A109011-500G A109011-5G
103823-210EA 5 USD
103823-210 77135-862 77135-864 77135-866 77135-868 77283-794
2-Hydroxybenzimidazole 98%
2-Hydroxybenzimidazole

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00127894
CAS Number: 615-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR