2-Hydroxy-6-methylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid

A211636-1G A211636-25G A211636-5G A211636-1MG A211636-250MG A211636-10G
103818-128EA 9 USD
103818-128 103848-822 77087-242 77087-244 77087-246 77261-070 77261-072 77748-317
2-Hydroxy-6-methylbenzoic acid 98%
2-Hydroxy-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 300.4 °C (760 mm Hg)
Melting Pt: 170…171 °C
Flash Pt: 149.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01194284
CAS Number: 567-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR