2-Hydroxy-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 6-(Trifluoromethyl)pyridin-2-ol

A381835-25G A381835-5G A381835-1G
103826-706EA 5 USD
103826-706 103837-740 76841-060 76841-062 77306-926
2-Hydroxy-6-(trifluoromethyl)pyridine 98%
2-Hydroxy-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD08061317
CAS Number: 34486-06-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR