5-Nitrosalicylic acid 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-nitrobenzoic acid

A498330-100G A498330-10G A498330-500G A498330-5G
103839-358EA 10 USD
103839-358 76774-468 76774-470 76774-472 76843-328
5-Nitrosalicylic acid 97% (by HPLC)
5-Nitrosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 228…230 °C
Flash Pt: 183.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00007338
CAS Number: 96-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR