2-Hydroxy-5-methylisophthalaldehyde 94%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

A131296-10G A131296-250MG A131296-1G A131296-5G
103801-558EA 18 USD
103801-558 77258-478 77258-480 77314-050 77314-052
2-Hydroxy-5-methylisophthalaldehyde 94%
2-Hydroxy-5-methylisophthalaldehyde

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.1600 g/mol
Melting Pt: 128…130 °C
Flash Pt: 111.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00016596
CAS Number: 7310-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR