5-Methylsalicylaldehyde 98%

Supplier: Ambeed
Warning

A132681-100G A132681-10G A132681-500G A132681-25G A132681-5G
76688-698EA 63 USD
76688-698 76688-700 76688-702 77298-572 77314-342
5-Methylsalicylaldehyde 98%
5-Methylsalicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
Boiling Pt: 217 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 85.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00274297
CAS Number: 613-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR