5-Methoxysalicylaldehyde 97%

Supplier: Ambeed
Warning

A120421-100G A120421-10G A120421-25G A120421-500G A120421-1G A120421-5G
103799-926EA 6 USD
103799-926 77246-076 77246-078 77246-080 77246-082 77297-756 77297-758
5-Methoxysalicylaldehyde 97%
5-Methoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
Boiling Pt: 247.5 °C (760 mm Hg)
Melting Pt: 4 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR