5-Iodosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-iodobenzaldehyde

A328577-1G A328577-25G A328577-5G A328577-100G A328577-10G A328577-250MG
103825-500EA 8 USD
103825-500 103836-454 76839-416 76839-418 76839-420 77141-906 77141-908 77141-910
5-Iodosalicylaldehyde 98%
5-Iodosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₅IO₂
MW: 248.0200 g/mol
Boiling Pt: 282 °C (760 mm Hg)
Melting Pt: 98…100 °C
Flash Pt: 124.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00723501
CAS Number: 1761-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR