2-Hydroxy-4-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A186069-1G A186069-100MG A186069-250MG A186069-25G A186069-5G
103815-908EA 20 USD
103815-908 103846-482 77084-130 77260-380 77260-382 77302-014 77302-016
2-Hydroxy-4-methylbenzonitrile 98%
2-Hydroxy-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.1500 g/mol
Boiling Pt: 286.1 °C (760 mm Hg)
Flash Pt: 126.797 °C
Storage Temperature: Ambient
MDL Number: MFCD02261941
CAS Number: 18495-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR