2-Hydroxy-5-nitro-4-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-methyl-5-nitropyridine, 4-Methyl-5-nitro-2-pyridone

A191405-25G A191405-100G A191405-10G A191405-500G A191405-1G A191405-5G
103816-370EA 6 USD
103816-370 103846-974 77084-810 77111-760 77111-762 77111-764 77302-258 77302-260
2-Hydroxy-5-nitro-4-picoline 97%
2-Hydroxy-5-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Boiling Pt: 292 °C (760 mm Hg)
Melting Pt: 186…190 °C (760 mm Hg)
Flash Pt: 130.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010690
CAS Number: 21901-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR