4-Methoxysalicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid

A183049-25G A183049-10G A183049-1G A183049-5G A183049-100G A183049-500G
103815-598EA 5 USD
103815-598 103846-162 77083-748 77274-168 77301-818 77301-820 77615-548 77748-192
4-Methoxysalicylic acid 98%
4-Methoxysalicylic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327
Melting Pt: 191
Flash Pt: 137.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00002450
CAS Number: 2237-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR