4,6-Dimethoxysalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4,6-dimethoxybenzaldehyde, 4,6-Dimethoxy-2-hydroxybenzaldehyde

A271155-1G A271155-25G A271155-5G A271155-100G A271155-100MG A271155-10G A271155-250MG
103813-038EA 9 USD
103813-038 103852-948 77092-722 77092-724 77092-726 77250-344 77250-346 77250-348 77250-350
4,6-Dimethoxysalicylaldehyde 98%
4,6-Dimethoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003328
CAS Number: 708-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR