2-Hydroxy-3-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A136793-250MG A136793-100MG
103802-354EA 17 USD
103802-354 77162-130 77891-952
2-Hydroxy-3-methylbenzonitrile 98%
2-Hydroxy-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO
Storage Temperature: Refrigerator
MDL Number: MFCD09836340
CAS Number: 13589-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR