2-Hydroxy-3-methoxy-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A270221-25G A270221-5G
103812-992EA 10 USD
103812-992 103852-898 77305-048 77305-050
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde 98%
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Boiling Pt: 344 °C (760 mm Hg)
Melting Pt: 140…142 °C (760 mm Hg)
Flash Pt: 161.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017033
CAS Number: 17028-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR