2-Hydroxy-1-naphthaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-α-naphthaldehyde

A176302-100G A176302-500G A176302-25G A176302-5G
103809-096EA 274 USD
103809-096 103845-452 76799-542 76799-544 77301-442 77301-444
2-Hydroxy-1-naphthaldehyde 98%
2-Hydroxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004005
CAS Number: 708-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR