2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1-(4-(2-hydroxyethoxy)phenyl)-2-methylpropan-1-one

A565451-25G A565451-100G A565451-10G A565451-500G
103839-908EA 13 USD
103839-908 76844-024 77005-220 77005-222 77005-224
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98%
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.2530 g/mol
Boiling Pt: 405.0±25.0 °C (760 mm Hg)
Melting Pt: 88…90 °C
Flash Pt: 155.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00085267
CAS Number: 106797-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR