2-Hydrazino-5-(trifluoromethyl)pyridine, HCl 95%

Supplier: Ambeed
Warning

A100254-250MG A100254-1G A100254-5G
103795-322EA 39 USD
103795-322 103821-032 77093-456 77296-614 77296-616
2-Hydrazino-5-(trifluoromethyl)pyridine, HCl 95%
2-Hydrazino-5-(trifluoromethyl)pyridine, HCl

Precaución: for Research Use Only

Formula: C₆H₇ClF₃N₃
MW: 213.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04966827
CAS Number: 1049744-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR