2-Hexylthiophene 98%

Supplier: Ambeed
Warning

A120509-10G A120509-25G A120509-5G A120509-100G
103799-948EA 5 USD
103799-948 77271-468 77271-470 77297-770 77747-876
2-Hexylthiophene 98%
2-Hexylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₆S
MW: 168.3 g/mol
Boiling Pt: 222 °C (760 mm Hg)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00022535
CAS Number: 18794-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR