2H-1,2,4-Triazole-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2H-1,2,4-triazole-3-carbonitrile

A186124-1G A186124-100G A186124-10G A186124-25G A186124-5G
103815-918EA 5 USD
103815-918 103846-492 77084-144 77287-410 77287-412 77287-414 77302-024
2H-1,2,4-Triazole-3-carbonitrile 97%
2H-1,2,4-Triazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₃H₂N₄
MW: 94.0700 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD11505654
CAS Number: 3641-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR