2-Furonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 2-Cyanofuran

A163728-25G A163728-5G A163728-1G
103805-760EA 7 USD
103805-760 103844-022 77300-672 77300-674 77320-132
2-Furonitrile 98%
2-Furonitrile

Precaución: for Research Use Only

Formula: C₅H₃NO
MW: 93.08 g/mol
Boiling Pt: 146…148 °C (760 mm Hg)
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003223
CAS Number: 617-90-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR