2-Furohydrazide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Furoic acid hydrazide, 2-Furoylhydrazine, 2-Furancarboxylic acid Hydrazide, 2-Furoic hydrazide, 2-Furoic acid Hydrazide, 2-Furancarboxylic acid hydrazide, 2-Furoic Hydrazide

A449735-5G A449735-1G
103827-782EA 5 USD
103827-782 103838-850 76842-666 77307-354
2-Furohydrazide 97%
2-Furohydrazide

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Melting Pt: 76…80 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS Number: 3326-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR