2-Fluoropyridine 98%

Supplier: Ambeed
Danger

A854596-25G A854596-10G
103841-248EA 5 USD
103841-248 76845-504 77105-942
2-Fluoropyridine 98%
2-Fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 125.9 °C (760 mm Hg)
Flash Pt: 28.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006224
CAS Number: 372-48-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR