2-Fluoro-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoropyridin-3-ol, 2-Fluoro-3-pyridinol

A192849-10G A192849-1G A192849-25G A192849-5G
103816-488EA 5 USD
103816-488 103847-098 76999-034 77084-984 77084-986 77084-988
2-Fluoro-3-hydroxypyridine 98%
2-Fluoro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
MW: 113.09 g/mol
Boiling Pt: 306.7 °C (760 mm Hg)
Melting Pt: 131…133 °C
Flash Pt: 139.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04112569
CAS Number: 174669-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR