3-Amino-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A167358-1G A167358-5G A167358-10G
103806-174EA 9 USD
103806-174 103844-434 77081-440 77081-442 77286-660
3-Amino-2-fluoropyridine 97%
3-Amino-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
Boiling Pt: 243.2 °C (760 mm Hg)
Flash Pt: 100.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095248
CAS Number: 1597-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR