2-Fluoronicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

A151273-100G A151273-25G A151273-5G
103804-240EA 5 USD
103804-240 103842-388 77095-818 77317-930 77317-932
2-Fluoronicotinic acid 97%
2-Fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 298.7 °C (760 mm Hg)
Melting Pt: 161…165 °C
Flash Pt: 122.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00040744
CAS Number: 393-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR