2-Fluoronaphthalene 98%

Supplier: Ambeed
Danger

A832635-1G A832635-25G A832635-100MG A832635-250MG A832635-5G
103841-174EA 145 USD
103841-174 76845-408 76845-410 77268-474 77268-476 77308-420
2-Fluoronaphthalene 98%
2-Fluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇F
MW: 146.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082618
CAS Number: 323-09-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR