2-Fluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoropyridine-4-carboxylic acid

A167062-100G A167062-10G A167062-500G A167062-1G A167062-25G A167062-5G
103806-124EA 5 USD
103806-124 103844-386 77096-342 77096-344 77096-346 77320-666 77320-668 77320-670
2-Fluoroisonicotinic acid 97%
2-Fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.10 g/mol
Boiling Pt: 396.6 °C (760 mm Hg)
Melting Pt: 200 °C
Flash Pt: 193.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02181194
CAS Number: 402-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR