2-Fluoroethyl p-Toluenesulfonate 97%

Supplier: Ambeed
Warning

A143495-100G A143495-10G A143495-250MG A143495-1G A143495-25G A143495-5G
103803-290EA 14 USD
103803-290 77258-910 77258-912 77258-914 77316-564 77316-566 77316-568
2-Fluoroethyl p-Toluenesulfonate 97%
2-Fluoroethyl p-Toluenesulfonate

Precaución: for Research Use Only

Formula: C₉H₁₁FO₃S
Boiling Pt: 335 °C (760 mm Hg)
Flash Pt: 156.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01658061
CAS Number: 383-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR