2-Fluorobenzene-1,4-diol 95%

Supplier: Ambeed
Warning

A111896-250MG A111896-1G A111896-25G
103797-920EA 37 USD
103797-920 103823-744 76821-882 77310-552 77885-031
2-Fluorobenzene-1,4-diol 95%
2-Fluorobenzene-1,4-diol

Precaución: for Research Use Only

Formula: C₆H₅FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00061106
CAS Number: 55660-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR