2-Fluoroadenine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-fluoropurine, 6-Amino-2-fluoro-7H-purine, 2-Fluoro-7H-purin-6-amine

A146219-100MG A146219-250MG A146219-25G A146219-1G A146219-5G
103803-646EA 13 USD
103803-646 103841-732 77285-942 77285-944 77299-506 77317-046 77317-048
2-Fluoroadenine 97%
2-Fluoroadenine

Precaución: for Research Use Only

Formula: C₅H₄FN₅
MW: 153.12 g/mol
Boiling Pt: 676.9 °C (760 mm Hg)
Melting Pt: >350 °C
Flash Pt: 363.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01632749
CAS Number: 700-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR