2-Fluoropropenoic acid 98%

Supplier: Ambeed
Danger

A101455-100MG A101455-10G A101455-250MG A101455-25G A101455-1G A101455-5G
103795-982EA 9 USD
103795-982 103821-666 77257-240 77257-242 77257-244 77257-246 77297-106 77297-108
2-Fluoropropenoic acid 98%
2-Fluoropropenoic acid

Precaución: for Research Use Only

Formula: C₃H₃FO₂
Boiling Pt: 181.3 °C (760 mm Hg)
Melting Pt: 51.5 °C
Flash Pt: 63.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03424474
CAS Number: 430-99-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR