2-Fluoro-6-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A176266-10G A176266-250MG A176266-25G A176266-5G
103807-126EA 13 USD
103807-126 103845-446 77111-210 77111-212 77301-438 77301-440
2-Fluoro-6-nitrobenzonitrile 95%
2-Fluoro-6-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
Boiling Pt: 302.9 °C (760 mm Hg)
Melting Pt: 68…71 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD08063904
CAS Number: 143306-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR